Merck
  • Design and biological evaluation of imidazo[1,2-a]pyridines as novel and potent ASK1 inhibitors.
Design and biological evaluation of imidazo[1,2-a]pyridines as novel and potent ASK1 inhibitors.
Bioorganic & medicinal chemistry letters (2012-11-14)
Yoshito Terao, Hideo Suzuki, Masato Yoshikawa, Hiroaki Yashiro, Shiro Takekawa, Yasushi Fujitani, Kengo Okada, Yoshihisa Inoue, Yoshio Yamamoto, Hideyuki Nakagawa, Shuhei Yao, Tomohiro Kawamoto, Osamu Uchikawa
ABSTRACT
Imidazo[1,2-a]pyridine derivatives were designed, synthesized, and evaluated as inhibitors of the apoptosis signal-regulating kinase 1 (ASK1). These were based on a benzothiazole derivative that was discovered from high-throughput screening of our compound library. As a result, we identified potent, selective, and orally bioavailable ASK1 inhibitors for wide range of therapeutic targets.
Sigma-Aldrich
Imidazo[1,2-a]pyridine, 99%