Merck
  • Transdermal granisetron: a guide to its use in preventing nausea and vomiting induced by chemotherapy.
Transdermal granisetron: a guide to its use in preventing nausea and vomiting induced by chemotherapy.
CNS drugs (2012-07-28)
Gillian M Keating, Sean T Duggan, Monique P Curran
ABSTRACT
Transdermal granisetron (Sancuso®) is effective in the prevention of nausea and vomiting in patients with cancer who are receiving moderately or highly emetogenic chemotherapy for 3-5 days. Transdermal granisetron is noninferior to oral granisetron in this indication, and is generally well tolerated in this indication. Thus, transdermal granisetron provides a convenient option for the prevention of chemotherapy-induced nausea and vomiting, with the potential to improve patient compliance.
Product Description
Granisetron hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Granisetron hydrochloride, ≥98% (HPLC), solid