Merck
  • Progress of bis(heteroaryl)piperazines (BHAPs) as non-nucleoside reverse transcriptase inhibitors (NNRTIs) against human immunodeficiency virus type 1 (HIV-1).
Progress of bis(heteroaryl)piperazines (BHAPs) as non-nucleoside reverse transcriptase inhibitors (NNRTIs) against human immunodeficiency virus type 1 (HIV-1).
Mini reviews in medicinal chemistry (2010-04-13)
Hui Xu
ABSTRACT
Since the first case of acquired immunodeficiency syndrome (AIDS) was reported in 1981, AIDS, as the global disease affecting 33.2 million people in 2007, has always been an unsolved problem worldwide. Reverse transcriptase (RT) is a crucial enzyme in the life cycle of human immunodeficiency virus type 1 (HIV-1), and thereby has been the prime drugs target for antiretroviral (ARV) therapy against AIDS. To date, two classes of RT inhibitors (RTIs), e.g., nucleoside reverse transcriptase inhibitors (NRTIs) and non-nucleoside reverse transcriptase inhibitors (NNRTIs), and a lot of compounds tested as RTIs have been described. To our knowledge, bis(heteroaryl)piperazines (BHAPs) have been considered as one class of promising NNRTIs, such as structurally and chemically related NNRTI delavirdine, which was approved by the U. S. Food and Drug Administration (FDA) for the treatment of HIV-1 infection in 1997. In this mini-review, we make attempts to report the progress of synthesis and structure-activity relationship (SAR) of BHAPs, in the meantime, the synergistic inhibition of HIV-1 replication by combining delavirdine with other HIV-1 inhibitors is also discussed. It will pave the way for the design and development of BHAPs as anti-HIV-1 agents in AIDS chemotherapy in the future.
Sigma-Aldrich
Delavirdine mesylate, ≥98% (HPLC)