Merck
  • Do social parasitic bumblebees use chemical weapons? (Hymenoptera, Apidae).
Do social parasitic bumblebees use chemical weapons? (Hymenoptera, Apidae).
Journal of comparative physiology. A, Neuroethology, sensory, neural, and behavioral physiology (2003-09-05)
B O Zimma, M Ayasse, J Tengö, F Ibarra, C Schulz, W Francke
ABSTRACT
The bumblebee Bombus (Psithyrus) norvegicus Sp.-Schn. is an obligate social parasite of B. (Pyrobombus) hypnorum L. Behavioural observations indicated that nest-invading B. norvegicus females may use allomones to defend themselves against attacking host workers. However, so far no defensive chemicals used by social parasitic bumblebee females have been identified. We analysed volatile constituents of the cuticular lipid profile of B. norvegicus females. Furthermore, we performed electrophysiological studies and behavioural experiments in order to identify possible chemical weapons. Coupled gas chromatography-electroantennography showed 15 compounds to trigger responses in antennae of the host workers. Using gas chromatography-mass spectrometry, the main compound among the cuticular volatiles of B. norvegicus females was found to be dodecyl acetate. A corresponding mixture of synthetic volatiles as well as pure dodecyl acetate showed a strong repellent effect on starved host workers. B. norvegicus females use dodecyl acetate to repel attacking B. hypnorum workers during nest usurpation and subsequently during colony development. Dodecyl acetate is the first repellent allomone identified in bumblebees.
Sigma-Aldrich
Dodecyl acetate, 97%
Sigma-Aldrich
Lauryl acetate, ≥98%, FG