Merck
  • Synthesis of N4-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-L-asparagine analogues. n-Butyramide, 3-chloropropionamide, 3-aminopropionamide, and isovaleramide analogues.
Synthesis of N4-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-L-asparagine analogues. n-Butyramide, 3-chloropropionamide, 3-aminopropionamide, and isovaleramide analogues.
Carbohydrate research (2001-06-12)
J J Kaylor, J M Risley
ABSTRACT
The syntheses of four analogues of N4-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-L-asparagine are described. Activated carboxylic acids were reacted with 2-acetamido-2-deoxy-beta-D-glucopyranosylamine. n-Butyric anhydride gave N-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-n-butyramide. 3-Chloropropionic anhydride was synthesized from 3-chloropropionic acid and gave N-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-3-chloropropionamide. Equilibration of the latter with ammonium bicarbonate gave N1-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-3-aminopropionamide. Succinimidyl isovalerate was synthesized and gave N-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-isovaleramide.
Sigma-Aldrich
Butyric anhydride, purum, ≥97.0% (NT)
Sigma-Aldrich
Butyramide, ≥98.0% (T)