Merck
  • Combining quantum mechanical ligand conformation analysis and protein modeling to elucidate GPCR-ligand binding modes.
Combining quantum mechanical ligand conformation analysis and protein modeling to elucidate GPCR-ligand binding modes.
ChemMedChem (2012-11-20)
Sabine Schultes, Harald Engelhardt, Luc Roumen, Obbe P Zuiderveld, Eric E J Haaksma, Iwan J P de Esch, Rob Leurs, Chris de Graaf
ABSTRACT
SAR beyond protein-ligand interactions: By combining structure-affinity relationships, protein-ligand modeling studies, and quantum mechanical calculations, we show that ligand conformational energies and basicity play critical roles in ligand binding to the histamine H4 receptor, a GPCR that plays a key role in inflammation.
Sigma-Aldrich
2-Aminopyrimidine, 97%