Merck
  • A rapid method for detection of fumonisins B1 and B2 in corn meal using Fourier transform near infrared (FT-NIR) spectroscopy implemented with integrating sphere.
A rapid method for detection of fumonisins B1 and B2 in corn meal using Fourier transform near infrared (FT-NIR) spectroscopy implemented with integrating sphere.
Food chemistry (2012-09-08)
B Gaspardo, S Del Zotto, E Torelli, S R Cividino, G Firrao, G Della Riccia, B Stefanon
ABSTRACT
Fourier transform near infrared (FT-NIR) spectroscopy is an analytical procedure generally used to detect organic compounds in food. In this work the ability to predict fumonisin B(1)+B(2) contents in corn meal using an FT-NIR spectrophotometer, equipped with an integration sphere, was assessed. A total of 143 corn meal samples were collected in Friuli Venezia Giulia Region (Italy) and used to define a 15 principal components regression model, applying partial least square regression algorithm with full cross validation as internal validation. External validation was performed to 25 unknown samples. Coefficients of correlation, root mean square error and standard error of calibration were 0.964, 0.630 and 0.632, respectively and the external validation confirmed a fair potential of the model in predicting FB(1)+FB(2) concentration. Results suggest that FT-NIR analysis is a suitable method to detect FB(1)+FB(2) in corn meal and to discriminate safe meals from those contaminated.
Product Description
Supelco
Fumonisin B1 solution, ~50 μg/mL in acetonitrile: water, analytical standard
Supelco
Fumonisin B1, reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Fumonisin B1 from Fusarium moniliforme, ≥98% (HPLC)