Merck
  • Minimalist probes for studying protein dynamics: thioamide quenching of selectively excitable fluorescent amino acids.
Minimalist probes for studying protein dynamics: thioamide quenching of selectively excitable fluorescent amino acids.
Journal of the American Chemical Society (2012-04-05)
Jacob M Goldberg, Lee C Speight, Mark W Fegley, E James Petersson
ABSTRACT
Fluorescent probe pairs that can be selectively excited in the presence of Trp and Tyr are of great utility in studying conformational changes in proteins. However, the size of these probe pairs can restrict their incorporation to small portions of a protein sequence where their effects on secondary and tertiary structure can be tolerated. Our findings show that a thioamide bond-a single atom substitution of the peptide backbone-can quench fluorophores that are red-shifted from intrinsic protein fluorescence, such as acridone. Using steady-state and fluorescence lifetime measurements, we further demonstrate that this quenching occurs through a dynamic electron-transfer mechanism. In a proof-of-principle experiment, we apply this technique to monitor unfolding in a model peptide system, the villin headpiece HP35 fragment. Thioamide analogues of the natural amino acids can be placed in a variety of locations in a protein sequence, allowing one to make a large number of measurements to model protein folding.
Sigma-Aldrich
9(10H)-Acridanone, 99%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
32324-250MG250 mg
Ships in 14 days - from Aldrich Partner.
€475.19
32324-500MG500 mg
Ships in 14 days - from Aldrich Partner.
€654.00
32324-1G1 g
Ships in 14 days - from Aldrich Partner.
€891.61
To order products, please contact your local dealer.