Merck
  • A straightforward one-pot multicomponent synthesis of polysubstituted pyrroles.
A straightforward one-pot multicomponent synthesis of polysubstituted pyrroles.
Chemical communications (Cambridge, England) (2011-05-13)
Xufeng Lin, Zhenjun Mao, Xixiang Dai, Ping Lu, Yanguang Wang
ABSTRACT
Polysubstituted pyrroles were efficiently synthesized in moderate yields by a one-pot multicomponent reaction starting from primary amines, ethyl glyoxalate and 2-bromoacetophenones in the presence of pyridine in refluxing acetonitrile. This methodology was utilized to synthesize a highly substituted benz[g]indole.
Product Description
Supelco
2-Bromoacetophenone, for GC derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
2-Bromoacetophenone, 98%