Merck
  • The influence of secondary interactions on complex stability and double proton transfer reaction in 2-[1H]-pyridone/2-hydroxypyridine dimers.
The influence of secondary interactions on complex stability and double proton transfer reaction in 2-[1H]-pyridone/2-hydroxypyridine dimers.
Journal of molecular modeling (2011-01-05)
Borys Ośmiałowski, Robert Dobosz
ABSTRACT
The 2-[1H]-pyridone/2-hydroxypyridine tautomeric pair and its 6-substituted complexes have been studied with the use of DFT(M05) method. The intermolecular interaction energy has been calculated and discussed in the light of secondary interaction concept. The attractive secondary interactions of O/NH and O/OH type and OH/NH and OH/OH repulsions have been analyzed in terms of stabilizing or destabilizing influence on intermolecular behavior. The transition states of the double proton transfer reaction have been found and the energy of activation has been determined. The activation energy of the proton transfer reaction, geometry of the complexes and transition states show NH(2) and/or OH groups influence the properties of complexes and transition states. The HOMA index of aromaticity was applied to describe the π-electron delocalization in the heterocyclic rings.
Product Description
Levetiracetam impurity C, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Hydroxypyridine, 97%