Merck
  • Highly selective recognition of carbenicillin via concerted interactions in 100% aqueous solution.
Highly selective recognition of carbenicillin via concerted interactions in 100% aqueous solution.
Chemical communications (Cambridge, England) (2010-04-10)
Lintao Zeng, Weimin Liu, Xiaoqing Zhuang, Jiasheng Wu, Pengfei Wang, Wenjun Zhang
ABSTRACT
A fluorescent probe (QA) based on quinine bearing two quaternary ammonium groups and a long hydrophobic chain was synthesized, and it showed highly selective recognition of carbenicillin (a typical beta-lactam antibiotic) in 100% aqueous solution via concerted interactions.
Sigma-Aldrich
Carbenicillin disodium salt, 89.0-100.5% anhydrous basis
Sigma-Aldrich
Carbenicillin disodium salt, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Carbenicillin disodium, meets USP testing specifications