Merck
  • The effect of laser wavelength on the Raman Spectra of phenanthrene, chrysene, and tetracene: implications for extra-terrestrial detection of polyaromatic hydrocarbons.
The effect of laser wavelength on the Raman Spectra of phenanthrene, chrysene, and tetracene: implications for extra-terrestrial detection of polyaromatic hydrocarbons.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2010-03-24)
A I Alajtal, H G M Edwards, M A Elbagerma, I J Scowen
ABSTRACT
Raman spectroscopy, with visible laser (514 and 633 nm) and near infrared (785 and 1064 nm) excitation, has been used to obtain high quality spectra of phenanthrene, chrysene, and tetracene. Samples with dimensions from a minimum size of 10 microm have been analyzed utilizing a Raman microprobe fitted with a charge-coupled device (CCD) array detector and a FT-Raman instrument. Fluorescence is observed for samples using visible 514, 633 and near infrared 785 nm excitation but most of the samples can be measured with a near infrared 1064 nm Nd:YAG laser.
Product Description
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chrysene, analytical standard
Chrysene, BCR®, certified reference material