Merck
  • Application of the margin of exposure (MoE) approach to substances in food that are genotoxic and carcinogenic: EXAMPLE 12: Sudan I (CAS No. 842-07-9).
Application of the margin of exposure (MoE) approach to substances in food that are genotoxic and carcinogenic: EXAMPLE 12: Sudan I (CAS No. 842-07-9).
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2010-02-02)
Myriam Coulet, Jean-Charles Leblanc, R Woodrow Setzer
ABSTRACT
Sudan I is generally considered mutagenic based on various in vitro and in vivo tests and is carcinogenic in the rat. Dose-response modelling of the data for hepatocellular adenomas in male rats gave a BMDL(10) of 7.3mg/kg-bw/day. Sudan I is an unauthorised substance that might be present in food intermittently. The great variability and uncertainties in the human exposure data which are country specific, depending on consumption patterns and methodology used, resulted in a large range of MOE values (from 30 to 2,000,000).
Product Description
Supelco
Sudan I, analytical standard
Sigma-Aldrich
Sudan I, Dye content ≥95 %