Merck
  • Drug-inducible remote control of gene expression by probiotic Escherichia coli Nissle 1917 in intestine, tumor and gall bladder of mice.
Drug-inducible remote control of gene expression by probiotic Escherichia coli Nissle 1917 in intestine, tumor and gall bladder of mice.
Microbes and infection (2009-08-12)
Holger Loessner, Sara Leschner, Anne Endmann, Kathrin Westphal, Kathrin Wolf, Katja Kochruebe, Tewfik Miloud, Josef Altenbuchner, Siegfried Weiss
ABSTRACT
The probiotic bacterium Escherichia coli Nissle 1917 (EcN) constitutes a prospective vector for delivering heterologous therapeutic molecules to treat several human disorders. To add versatility to this carrier system, bacteria should be equipped with expression modules that can be regulated deliberately in a temporal and quantitative manner. This approach is called in vivo remote control (IVRC) of bacterial vectors. Here, we have evaluated promoters P(araBAD), P(rhaBAD) and P(tet), which can be induced with L-arabinose, L-rhamnose or anhydrotetracycline, respectively. EcN harboring promoter constructs with luciferase as reporter gene were administered either orally to healthy mice or intravenously to tumor bearing animals. Subsequent to bacterial colonization of tissues, inducer substances were administered via the oral or systemic route. By use of in vivo bioluminescence imaging, the time course of reporter gene expression was analyzed. Each promoter displayed a specific in vivo induction profile depending on the niche of bacterial residence and the route of inducer administration. Importantly, we also observed colonization of gall bladders of mice when EcN was administered systemically at high doses. Bacteria in this anatomical compartment remained accessible to remote control of bacterial gene expression.
Product Description
Supelco
4-Epianhydrotetracycline hydrochloride, VETRANAL®, analytical standard
Supelco
Anhydrotetracycline hydrochloride, VETRANAL®, analytical standard
Supelco
Anhydrotetracycline hydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland