Merck
  • Synthesis and cytotoxic activity of a glucuronylated prodrug of nornitrogen mustard.
Synthesis and cytotoxic activity of a glucuronylated prodrug of nornitrogen mustard.
Bioorganic & medicinal chemistry letters (2000-09-02)
S Papot, D Combaud, K Bosslet, M Gerken, J Czech, J P Gesson
ABSTRACT
A new glucuronylated prodrug of nornitrogen mustard, incorporating the same spacer group as the doxorubicin prodrug HMR 1826, has been prepared. Upon exposure to E. coli beta-glucuronidase, fast hydrolysis occurs but a lower cytotoxicity against LoVo cancer cells is observed compared to the nornitrogen mustard alone. This is explained by cyclization of the intermediate carbamic acid to the inactive chloroethyl oxazolidinone.
Sigma-Aldrich
Bis(2-chloroethyl)amine hydrochloride, 98%