Merck
  • Tau forms synaptic nano-biomolecular condensates controlling the dynamic clustering of recycling synaptic vesicles.
Tau forms synaptic nano-biomolecular condensates controlling the dynamic clustering of recycling synaptic vesicles.
Nature communications (2023-11-11)
Shanley F Longfield, Mahdie Mollazade, Tristan P Wallis, Rachel S Gormal, Merja Joensuu, Jesse R Wark, Ashley J van Waardenberg, Christopher Small, Mark E Graham, Frédéric A Meunier, Ramón Martínez-Mármol
ABSTRACT
Neuronal communication relies on the release of neurotransmitters from various populations of synaptic vesicles. Despite displaying vastly different release probabilities and mobilities, the reserve and recycling pool of vesicles co-exist within a single cluster suggesting that small synaptic biomolecular condensates could regulate their nanoscale distribution. Here, we performed a large-scale activity-dependent phosphoproteome analysis of hippocampal neurons in vitro and identified Tau as a highly phosphorylated and disordered candidate protein. Single-molecule super-resolution microscopy revealed that Tau undergoes liquid-liquid phase separation to generate presynaptic nanoclusters whose density and number are regulated by activity. This activity-dependent diffusion process allows Tau to translocate into the presynapse where it forms biomolecular condensates, to selectively control the mobility of recycling vesicles. Tau, therefore, forms presynaptic nano-biomolecular condensates that regulate the nanoscale organization of synaptic vesicles in an activity-dependent manner.
Sigma-Aldrich
Anti-Tau, a.a. 210-241 Antibody, clone Tau-5, clone TAU-5, from mouse
Sigma-Aldrich
Anti-Green Fluorescent Protein Antibody, Chemicon®, from chicken