Merck
  • Identification of RNA helicases with unwinding activity on angiogenin-processed tRNAs.
Identification of RNA helicases with unwinding activity on angiogenin-processed tRNAs.
Nucleic acids research (2023-02-01)
Aleksej Drino, Lisa König, Charlotte Capitanchik, Nasim Sanadgol, Eva Janisiw, Tom Rappol, Elisa Vilardo, Matthias R Schaefer
ABSTRACT
Stress-induced tRNA fragmentation upon environmental insult is a conserved cellular process catalysed by endonucleolytic activities targeting mature tRNAs. The resulting tRNA-derived small RNAs (tsRNAs) have been implicated in various biological processes that impact cell-to-cell signalling, cell survival as well as gene expression regulation during embryonic development. However, how endonuclease-targeted tRNAs give rise to individual and potentially biologically active tsRNAs remains poorly understood. Here, we report on the in vivo identification of proteins associated with stress-induced tsRNAs-containing protein complexes, which, together with a 'tracer tRNA' assay, were used to uncover enzymatic activities that can bind and process specific endonuclease-targeted tRNAs in vitro. Among those, we identified conserved ATP-dependent RNA helicases which can robustly separate tRNAs with endonuclease-mediated 'nicks' in their anticodon loops. These findings shed light on the existence of cellular pathways dedicated to producing individual tsRNAs after stress-induced tRNA hydrolysis, which adds to our understanding as to how tRNA fragmentation and the resulting tsRNAs might exert physiological impact.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Angiogenin Antibody, clone 26–2F (Azide Free), clone 26-2F, from mouse
Pricing and availability is not currently available.