Merck
  • IQCN disruption causes fertilization failure and male infertility due to manchette assembly defect.
IQCN disruption causes fertilization failure and male infertility due to manchette assembly defect.
EMBO molecular medicine (2022-11-03)
Jing Dai, Qi Li, Qinwei Zhou, Shen Zhang, Junru Chen, Yize Wang, Jing Guo, Yifan Gu, Fei Gong, Yueqiu Tan, Guangxiu Lu, Wei Zheng, Ge Lin
ABSTRACT
Total fertilization failure (TFF) is an important cause of infertility; however, the genetic basis of TFF caused by male factors remains to be clarified. In this study, whole-exome sequencing was firstly used to screen for genetic causes of TFF after intracytoplasmic sperm injection (ICSI), and homozygous variants in the novel gene IQ motif-containing N (IQCN) were identified in two affected individuals with abnormal acrosome structures. Then, Iqcn-knockout mice were generated by CRISPR-Cas9 technology and showed that the knockout male mice resembled the human phenotypes. Additionally, we found that IQCN regulates microtubule nucleation during manchette assembly via calmodulin and related calmodulin-binding proteins, which resulted in head deformity with aberrant oocyte activation factor PLCζ. Fortunately, ICSI with assisted oocyte activation can overcome IQCN-associate TFF and male infertility. Thus, our study firstly identified the function of IQCN, highlights the relationship between the manchette assembly and fertilization, and provides a genetic marker and a therapeutic option for male-source TFF.
Sigma-Aldrich
Anti-α-Tubulin−FITC antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
W-7, Hydrochloride, A cell-permeable and reversible calmodulin antagonist that inhibits myosin light chain kinase (IC₅₀ = 51 µM) and Ca2+-calmodulin-dependent phosphodiesterase (IC₅₀ = 28 µM).
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Lectin from Arachis hypogaea (peanut), FITC conjugate, lyophilized powder