Merck
  • Efficacy and safety of cefditoren pivoxil for exacerbations of chronic obstructive pulmonary disease: A prospective multicenter interventional study.
Efficacy and safety of cefditoren pivoxil for exacerbations of chronic obstructive pulmonary disease: A prospective multicenter interventional study.
Journal of infection and chemotherapy : official journal of the Japan Society of Chemotherapy (2019-04-16)
Taiga Miyazaki, Kiyoyasu Fukushima, Kohji Hashiguchi, Yuichi Inoue, Tomo Mihara, Toyomitsu Sawai, Naofumi Suyama, Tsutomu Kobayashi, Akira Kondo, Yuichi Fukuda, Yosuke Harada, Eisuke Sasaki, Norihito Kaku, Shotaro Ide, Takahiro Takazono, Tomomi Saijo, Kosuke Kosai, Yoshitomo Morinaga, Shigeki Nakamura, Kazuko Yamamoto, Yoshifumi Imamura, Koichi Izumikawa, Katsunori Yanagihara, Shigeru Kohno, Hiroshi Mukae
ABSTRACT
Oral antibiotic therapy for patients with acute exacerbations of chronic obstructive pulmonary disease (COPD) usually involves an aminopenicillin with clavulanic acid, a macrolide, or a quinolone. To date, however, the clinical efficacy and safety of the oral cephalosporin cefditoren pivoxil has not been evaluated in Japanese patients with acute exacerbations of COPD. We conducted a prospective, multicenter, single arm, interventional study from January 2013 to March 2017 to determine the efficacy and safety of oral administration of 200 mg cefditoren pivoxil three times daily for 7 days in a cohort of 29 eligible patients from 15 hospitals. The mean age (SD) of participants was 73.1 (8.1) years and 28 had a smoking history (the mean [SD] of smoking index, 1426.7 [931.7]). The primary efficacy endpoint was clinical response (cure rate) at test of cure, which was set at 5-10 days after treatment ceased. Of the 23 patients finally analyzed, cure was achieved in 15 (65.2%), while 8 (34.8%) remained uncured. Previous experience of acute exacerbations significantly affected the cure rate: none of the three patients who had at least two prior exacerbations were cured, while 15 of the 20 patients with one or fewer prior exacerbations were cured (p = 0.032). The microbiological eradication rate was 88.9% at test of cure. During treatment, mild pneumonia was reported as an adverse event in one patient (3.4%) but resolved within 10 days of onset. We conclude that cefditoren pivoxil represents a viable alternative for antibiotic therapy in patients with few prior exacerbations.
Sigma-Aldrich
Cefditoren pivoxil, ≥98% (HPLC)
Pricing and availability is not currently available.