Merck
  • Integrated regulation of PKA by fast and slow neurotransmission in the nucleus accumbens controls plasticity and stress responses.
Integrated regulation of PKA by fast and slow neurotransmission in the nucleus accumbens controls plasticity and stress responses.
The Journal of biological chemistry (2022-07-15)
Rachel Thomas, Adan Hernandez, David R Benavides, Wei Li, Chunfeng Tan, Alan Umfress, Florian Plattner, Ayanabha Chakraborti, Lucas Pozzo-Miller, Susan S Taylor, James A Bibb
ABSTRACT
Cortical glutamate and midbrain dopamine neurotransmission converge to mediate striatum-dependent behaviors, while maladaptations in striatal circuitry contribute to mental disorders. However, the crosstalk between glutamate and dopamine signaling has not been entirely elucidated. Here we uncover a molecular mechanism by which glutamatergic and dopaminergic signaling integrate to regulate cAMP-dependent protein kinase (PKA) via phosphorylation of the PKA regulatory subunit, RIIβ. Using a combination of biochemical, pharmacological, neurophysiological, and behavioral approaches, we find that glutamate-dependent reduction in cyclin-dependent kinase 5 (Cdk5)-dependent RIIβ phosphorylation alters the PKA holoenzyme autoinhibitory state to increase PKA signaling in response to dopamine. Furthermore, we show that disruption of RIIβ phosphorylation by Cdk5 enhances cortico-ventral striatal synaptic plasticity. In addition, we demonstrate that acute and chronic stress in rats inversely modulate RIIβ phosphorylation and ventral striatal infusion of a small interfering peptide that selectively targets RIIβ regulation by Cdk5 improves behavioral response to stress. We propose this new signaling mechanism integrating ventral striatal glutamate and dopamine neurotransmission is important to brain function, may contribute to neuropsychiatric conditions, and serves as a possible target for the development of novel therapeutics for stress-related disorders.
Sigma-Aldrich
Anti-GluR1 Antibody, Recombinant, rabbit monoclonal, Upstate®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody (rabbit), from rabbit, purified by affinity chromatography