Merck
  • Cultured Epidermal Autografts from Clinically Revertant Skin as a Potential Wound Treatment for Recessive Dystrophic Epidermolysis Bullosa.
Cultured Epidermal Autografts from Clinically Revertant Skin as a Potential Wound Treatment for Recessive Dystrophic Epidermolysis Bullosa.
The Journal of investigative dermatology (2019-05-06)
Wakana Matsumura, Yasuyuki Fujita, Satoru Shinkuma, Shotaro Suzuki, Saki Yokoshiki, Hideki Goto, Hiroshi Hayashi, Kota Ono, Masukazu Inoie, Shota Takashima, Chihiro Nakayama, Toshifumi Nomura, Hideki Nakamura, Riichiro Abe, Norihiro Sato, Hiroshi Shimizu
ABSTRACT
Inherited skin disorders have been reported recently to have sporadic normal-looking areas, where a portion of the keratinocytes have recovered from causative gene mutations (revertant mosaicism). We observed a case of recessive dystrophic epidermolysis bullosa treated with cultured epidermal autografts (CEAs), whose CEA-grafted site remained epithelized for 16 years. We proved that the CEA product and the grafted area included cells with revertant mosaicism. Based on these findings, we conducted an investigator-initiated clinical trial of CEAs from clinically revertant skin for recessive dystrophic epidermolysis bullosa. The donor sites were analyzed by genetic analysis, immunofluorescence, electron microscopy, and quantification of the reverted mRNA with deep sequencing. The primary endpoint was the ulcer epithelization rate per patient at 4 weeks after the last CEA application. Three patients with recessive dystrophic epidermolysis bullosa with 8 ulcers were enrolled, and the epithelization rate for each patient at the primary endpoint was 87.7%, 100%, and 57.0%, respectively. The clinical effects were found to persist for at least 76 weeks after CEA transplantation. One of the three patients had apparent revertant mosaicism in the donor skin and in the post-transplanted area. CEAs from clinically normal skin are a potentially well-tolerated treatment for recessive dystrophic epidermolysis bullosa.
Sigma-Aldrich
Anti-Collagen Type VII Antibody, CT, clone LH7.2, ascites fluid, clone LH7.2, Chemicon®