Merck
  • ERK signaling mediates resistance to immunomodulatory drugs in the bone marrow microenvironment.
ERK signaling mediates resistance to immunomodulatory drugs in the bone marrow microenvironment.
Science advances (2021-06-06)
Jiye Liu, Teru Hideshima, Lijie Xing, Su Wang, Wenrong Zhou, Mehmet K Samur, Tomasz Sewastianik, Daisuke Ogiya, Gang An, Shaobing Gao, Li Yang, Tong Ji, Giada Bianchi, Kenneth Wen, Yu-Tzu Tai, Nikhil Munshi, Paul Richardson, Ruben Carrasco, Yong Cang, Kenneth C Anderson
ABSTRACT
Immunomodulatory drugs (IMiDs) have markedly improved patient outcome in multiple myeloma (MM); however, resistance to IMiDs commonly underlies relapse of disease. Here, we identify that tumor necrosis factor (TNF) receptor-associated factor 2 (TRAF2) knockdown (KD)/knockout (KO) in MM cells mediates IMiD resistance via activation of noncanonical nuclear factor κB (NF-κB) and extracellular signal-regulated kinase (ERK) signaling. Within MM bone marrow (BM) stromal cell supernatants, TNF-α induces proteasomal degradation of TRAF2, noncanonical NF-κB, and downstream ERK signaling in MM cells, whereas interleukin-6 directly triggers ERK activation. RNA sequencing of MM patient samples shows nearly universal ERK pathway activation at relapse on lenalidomide maintenance therapy, confirming its clinical relevance. Combination MEK inhibitor treatment restores IMiD sensitivity of TRAF2 KO cells both in vitro and in vivo. Our studies provide the framework for clinical trials of MEK inhibitors to overcome IMiD resistance in the BM microenvironment and improve patient outcome in MM.
Product Description
Millipore
Millipore® Steriflip® Vacuum Tube Top Filter, pore size 0.45 μm, PVDF membrane, funnel capacity 50 mL, pack of 25 ea