Merck
  • Acute inflammation down-regulates alpha-synuclein expression in enteric neurons.
Acute inflammation down-regulates alpha-synuclein expression in enteric neurons.
Journal of neurochemistry (2018-12-28)
Alice Prigent, Jacques Gonzales, Tony Durand, Catherine Le Berre-Scoul, Malvyne Rolli-Derkinderen, Michel Neunlist, Pascal Derkinderen
ABSTRACT
The protein alpha-synuclein whose expression is strongly implicated in Parkinson's disease (PD) is not only expressed in the CNS but also in the enteric nervous system (ENS). The growing body of evidence suggesting that gastrointestinal inflammation is involved in the development of PD led us to investigate the effects of inflammation on alpha-synuclein expression in primary culture of rat ENS and in mice with dextran sulfate sodium-induced colitis. Using western blot and qPCR, we found that both lipopolysaccharide and a combination of tumor necrosis factor-α and interleukin 1-β decreased the expression levels of alpha-synuclein in primary culture of rat ENS, an effect that was prevented in the presence of the p38 inhibitors SB203580 and BIRB 796. Lipopolysaccharide and tumor necrosis factor-α/interleukin 1-β had no effect on alpha-synuclein expression in primary culture of rat CNS and in human erythroid leukemia cells. In mice, acute but not chronic dextran sulfate sodium-induced colitis was associated with a decreased expression of colonic alpha-synuclein. As a whole, our findings indicate that acute inflammatory insults down-regulate alpha-synuclein expression in the ENS via a p38 pathway. They provide new insights into the widely discussed concepts of alpha-synuclein expression and aggregation in the ENS in PD and raise issues about the possible role of gastrointestinal inflammation in the development of PD. OPEN SCIENCE BADGES: This article has received a badge for *Open Materials* because it provided all relevant information to reproduce the study in the manuscript. The complete Open Science Disclosure form for this article can be found at the end of the article. More information about the Open Practices badges can be found at https://cos.io/our-services/open-science-badges/.
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
TWEEN® 20, viscous liquid
Millipore
RIPA Lysis Buffer, 10X, 100 mL RIPA Lysis Buffer, 10X for Immunoprecipitation & Western Blotting.
Sigma-Aldrich
SB 203580, Hydrochloride, SB 203580, Hydrochloride, CAS 869185-85-3, is a water-soluble form of SB 203580. A highly potent, selective, ATP-competitive inhibitor of p38 MAP Kinase (IC₅₀ = 34 nM in vitro, 600 nM in cells).
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Tris Buffered Saline, 10 ×, solution
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture