Merck
  • Epithelial-to-mesenchymal transition in the development and progression of adenocarcinoma and squamous cell carcinoma of the lung.
Epithelial-to-mesenchymal transition in the development and progression of adenocarcinoma and squamous cell carcinoma of the lung.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2009-03-10)
Ludmila Prudkin, Diane D Liu, Natalie C Ozburn, Menghong Sun, Carmen Behrens, Ximing Tang, Kathlynn C Brown, B Nebiyou Bekele, Cesar Moran, Ignacio I Wistuba
ABSTRACT
Epithelial-to-mesenchymal transition is a process in which cells undergo a developmental switch from an epithelial to a mesenchymal phenotype. We investigated the role of this phenomenon in the pathogenesis and progression of adenocarcinoma and squamous cell carcinoma of the lung. Archived tissue from primary tumors (n=325), brain metastases (n=48) and adjacent bronchial epithelial specimens (n=192) were analyzed for immunohistochemical expression by image analysis of E-cadherin, N-cadherin, integrin-alpha v beta 6, vimentin, and matrix metalloproteinase-9. The findings were compared with the patients' clinicopathologic features. High expression of the epithelial-to-mesenchymal transition phenotype (low E-cadherin and high N-cadherin, integrin-alpha v beta 6, vimentin, and matrix metalloproteinase-9) was found in most lung tumors examined, and the expression pattern varied according to the tumor histologic type. Low E-cadherin membrane and high N-cadherin cytoplasmic expression were significantly more common in squamous cell carcinoma than in adenocarcinoma (P=0.002 and 0.005, respectively). Dysplastic lesions had significantly lower expression of the epithelial-to-mesenchymal transition phenotype than the squamous cell carcinomas, and integrin-alpha v beta 6 membrane expression increased stepwise according to the histopathologic severity. Brain metastases had decreased epithelial-to-mesenchymal transition expression compared with primary tumors. Brain metastases had significantly lower integrin-alpha v beta 6 membrane (P=0.04), N-cadherin membrane, and cytoplasm (P<0.0002) expression than the primary tumors. The epithelial-to-mesenchymal transition phenotype is commonly expressed in primary squamous cell carcinoma and adenocarcinoma of the lung; this expression occurs early in the pathogenesis of squamous cell carcinoma. Brain metastases showed characteristics of reversed mesenchymal-to-epithelial transition. Our findings suggest that epithelial-to-mesenchymal transition is a potential target for lung cancer chemoprevention and therapy.
Sigma-Aldrich
Anti-Integrin β₆ Mouse mAb (442.5C4), liquid, clone 442.5C4, Calbiochem®