Merck
  • Synthesis of asymmetrical bispyridinium compounds bearing cyano-moiety and evaluation of their reactivation activity against tabun and paraoxon-inhibited acetylcholinesterase.
Synthesis of asymmetrical bispyridinium compounds bearing cyano-moiety and evaluation of their reactivation activity against tabun and paraoxon-inhibited acetylcholinesterase.
Bioorganic & medicinal chemistry letters (2006-08-29)
Kamil Musilek, Ondrej Holas, Kamil Kuca, Daniel Jun, Vlastimil Dohnal, Martin Dolezal
ABSTRACT
Three asymmetrical AChE reactivators with cyano-moiety and propane linker were synthesized using modification of currently known synthetic pathways. Their potency to reactivate AChE inhibited by nerve agent tabun and insecticide paraoxon was tested in vitro and compared to pralidoxime, HI-6, obidoxime, K027, and K048. According to the results, three compounds seem to be promising against paraoxon-inhibited AChE. Better results were obtained for bisquaternary substances at least with one oxime group in position four. None of tested substances was able to satisfactorily reactivate tabun-inhibited AChE at concentration applicable for in vivo experiments.
Sigma-Aldrich
Pyridine-2-aldoxime methochloride