Merck
  • CHIR-258: a potent inhibitor of FLT3 kinase in experimental tumor xenograft models of human acute myelogenous leukemia.
CHIR-258: a potent inhibitor of FLT3 kinase in experimental tumor xenograft models of human acute myelogenous leukemia.
Clinical cancer research : an official journal of the American Association for Cancer Research (2005-07-22)
Daniel E Lopes de Menezes, Jing Peng, Evelyn N Garrett, Sharianne G Louie, Sang H Lee, Marion Wiesmann, Yan Tang, Lee Shephard, Cheryl Goldbeck, Yoko Oei, Helen Ye, Sharon L Aukerman, Carla Heise
ABSTRACT
Fms-like tyrosine kinase 3 (FLT3) encodes a receptor tyrosine kinase (RTK) for which activating mutations have been identified in a proportion of acute myelogenous leukemia (AML) patients and associated with poor clinical prognosis. Given the relevance of FLT3 mutations in AML, we investigated the activity of CHIR-258, an orally active, multitargeted small molecule, with potent activity against FLT3 kinase and class III, IV, and V RTKs involved in endothelial and tumor cell proliferation in AML models. CHIR-258 was tested on two human leukemic cell lines in vitro and in vivo with differing FLT3 mutational status [MV4;11 cells express FLT3 internal tandem duplications (ITD) versus RS4;11 cells with wild-type (WT) FLT3]. Antiproliferative activity of CHIR-258 against MV4;11 was approximately 24-fold greater compared with RS4;11, indicating more potent inhibition against cells with constitutively activated FLT3 ITD. Dose-dependent down modulation of receptor phosphorylation and downstream signaling [signal transducer and activator of transcription 5 (STAT5) and extracellular signal-regulated kinase (ERK)/mitogen-activated protein kinase] in MV4;11 cells with CHIR-258 confirmed the molecular mechanism of action. Target modulation of phospho-FLT3, phospho-STAT5, and phospho-ERK in MV4;11 tumors was achieved at biologically active doses of CHIR-258. Tumor regressions and eradication of AML cells from the bone marrow were shown in s.c. and bone marrow engraftment leukemic xenograft models. Tumor responses were characterized by decreased cellular proliferation and positive immunohistochemical staining for active caspase-3 and cleaved poly(ADP-ribose) polymerase, suggesting cell death was mediated in part via apoptosis. Our data indicate that CHIR-258 may be an effective therapy in FLT3-associated AML and warrants clinical trials.
Sigma-Aldrich
Dovitinib, ≥98% (HPLC)
Pricing and availability is not currently available.