Merck
  • Flavin adenine dinucleotide ameliorates hypertensive vascular remodeling via activating short chain acyl-CoA dehydrogenase.
Flavin adenine dinucleotide ameliorates hypertensive vascular remodeling via activating short chain acyl-CoA dehydrogenase.
Life sciences (2020-08-01)
Xiaoyi Zhong, Zhichao Ma, Yongshao Su, Zhonghong Li, Yingqin Liao, Xuediao Pan, Linquan Zang, Sigui Zhou
ABSTRACT
Flavin adenine dinucleotide (FAD), participates in fatty acid β oxidation as a cofactor, which has been confirmed to enhance SCAD activity and expression. However, the role of FAD on hypertensive vascular remodeling is unclear. In this study, we investigated the underlying mechanisms of FAD on vascular remodeling and endothelial homeostasis. Morphological examination of vascular remodeling were analyzed with hematoxylin and eosin (HE) staining, Verhoeff's Van Gieson (EVG) staing, Dihydroethidium (DHE) staining and Sirius red staining. HUVECs apoptotic rate was detected by flow cytometry and HUVECs reactive oxygen species (ROS) was detected by DHE-probe. Enzymatic reactions were used to detect SCAD enzyme activity. The protein level was detected by Western Blots, the mRNA level was detected by quantitative real-time PCR. In vivo experiments, FAD significantly decreased blood pressure and ameliorated vascular remodeling by increasing SCAD expression, Nitric Oxide (NO) production and reducing ROS production. In vitro experiments, FAD protected against the tBHP induced injury in HUVEC, by increasing the activity of SCAD, increasing the elimination of free fatty acid (FFA), scavenging ROS, reducing apoptotic rate, thereby improving endothelial cell function. FAD has a new possibility for preventing and treating hypertensive vascular remodeling.
Sigma-Aldrich
Flavin adenine dinucleotide disodium salt hydrate, ≥95% (HPLC), powder