Merck
  • Responses of AcidobacteriaGranulicella sp. WH15 to High Carbon Revealed by Integrated Omics Analyses.
Responses of AcidobacteriaGranulicella sp. WH15 to High Carbon Revealed by Integrated Omics Analyses.
Microorganisms (2020-02-16)
Ohana Y A Costa, Marcelo M Zerillo, Daniela Zühlke, Anna M Kielak, Agata Pijl, Katharina Riedel, Eiko E Kuramae
ABSTRACT
The phylum Acidobacteria is widely distributed in soils, but few representatives have been cultured. In general, Acidobacteria are oligotrophs and exhibit slow growth under laboratory conditions. We sequenced the genome of Granulicella sp. WH15, a strain obtained from decaying wood, and determined the bacterial transcriptome and proteome under growth in poor medium with a low or high concentration of sugar. We detected the presence of 217 carbohydrate-associated enzymes in the genome of strain WH15. Integrated analysis of the transcriptomic and proteomic profiles showed that high sugar triggered a stress response. As part of this response, transcripts related to cell wall stress, such as sigma factor σW and toxin-antitoxin (TA) systems, were upregulated, as were several proteins involved in detoxification and repair, including MdtA and OprM. KEGG metabolic pathway analysis indicated the repression of carbon metabolism (especially the pentose phosphate pathway) and the reduction of protein synthesis, carbohydrate metabolism, and cell division, suggesting the arrest of cell activity and growth. In summary, the stress response of Granulicella sp. WH15 induced by the presence of a high sugar concentration in the medium resulted in the intensification of secretion functions to eliminate toxic compounds and the reallocation of resources to cell maintenance instead of growth.
Product Description
Millipore
Tryptone enzymatic digest from casein, for biotechnological purposes
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG