Merck
Regulation of NF-κB by PML and PML-RARα.
Scientific reports (2017-03-21)
Abrar Ahmed, Xiaochun Wan, Izaskun Mitxitorena, Andrew J Lindsay, Pier Paolo Pandolfi, Mary W McCaffrey, Karen Keeshan, Youhai H Chen, Ruaidhrí J Carmody
ABSTRACT
Promyelocytic Leukemia (PML) is a nuclear protein that forms sub-nuclear structures termed nuclear bodies associated with transcriptionally active genomic regions. PML is a tumour suppressor and regulator of cell differentiation. We demonstrate that PML promotes TNFα-induced transcriptional responses by promoting NF-κB activity. TNFα-treated PML-/- cells show normal IκBα degradation and NF-κB nuclear translocation but significantly reduced NF-κB DNA binding and phosphorylation of NF-κB p65. We also demonstrate that the PML retinoic acid receptor-α (PML-RARα) oncofusion protein, which causes acute promyelocytic leukemia, inhibits TNFα induced gene expression and phosphorylation of NF-κB. This study establishes PML as an important regulator of NF-κB and demonstrates that PML-RARα dysregulates NF-κB.
Sigma-Aldrich
Anti-PML Antibody, clone 36.1-104, clone 36.1-104, Upstate®, from mouse