Merck
  • Physiological and molecular characterisation of cadmium stress in Schmidtea mediterranea.
Physiological and molecular characterisation of cadmium stress in Schmidtea mediterranea.
The International journal of developmental biology (2012-03-28)
Michelle Plusquin, An-Sofie Stevens, Frank Van Belleghem, Olivier Degheselle, Andromeda Van Roten, Jessica Vroonen, Ronny Blust, Ann Cuypers, Tom Artois, Karen Smeets
ABSTRACT
The planarian Schmidtea mediterranea is a well-studied model organism for developmental research, because of its stem cell system. This characteristic also provides a unique opportunity to study stress management and the effect of stress on stem cells. In this study, we characterised the stress signature at different levels of biological organization. The carcinogenic metal cadmium was used as a model chemical stressor. We focused on stem cell activity and its interaction with other known stress parameters. Here, we have found that S. mediterranea is able to cope with high internal levels of cadmium. At endpoints such as size and mobility, cadmium-related stress effects were detected but all of these responses were transient. Correspondingly, cadmium exposure led to an elevated mitotic activity of the neoblasts, at the same time points when the other responses disappeared. At the molecular level, we observed redox-related responses that can be linked with both repair as well as proliferation mechanisms. Together, our results suggest that these animals have a high plasticity. The induction of stem cell activity may underlie this 'restoring' effect, although a carcinogenic outcome after longer exposure times cannot be excluded.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, biotin conjugate, Upstate®, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Goat Anti-Rabbit IgG (H+L) Antibody, Rhodamine conjugate, Upstate®, from goat