Merck
  • Klotho inhibits angiotensin II-induced cardiac hypertrophy, fibrosis, and dysfunction in mice through suppression of transforming growth factor-β1 signaling pathway.
Klotho inhibits angiotensin II-induced cardiac hypertrophy, fibrosis, and dysfunction in mice through suppression of transforming growth factor-β1 signaling pathway.
European journal of pharmacology (2019-07-22)
Jieqiong Ding, Qiong Tang, Binhua Luo, Lijun Zhang, Li Lin, Lu Han, Miaomiao Hao, Mingyue Li, Liangzhu Yu, Mincai Li
ABSTRACT
Recent studies have revealed critical roles of transforming growth factor-β1 (TGF-β1) and microRNA-132 (miR-132), a downstream mediator of TGF-β1, in the pathogenesis of cardiac remodeling. In this study, we tested whether the antiaging protein klotho modifies angiotensin II (Ang II)-induced cardiac remodeling through regulating TGF-β1-miR-132 axis. We found that both klotho and the TGF-β1 inhibitor LY364947 significantly inhibited cardiac hypertrophy, fibrosis, and dysfunction in Ang II-infused mice, as evidenced by the ratios of heart weight to body weight (HW/BW), heart weight to tibial length (HW/TL), cardiomyocyte cross-sectional area, fibrotic area, and expression of prohypertrophic genes (ANP, β-MHC) and fibrotic marker genes (α-SMA, collagen I), echocardiographic parameters. Meanwhile, klotho also significantly inhibited Ang II-induced protein expression of TGF-β1 and phosphorylated Smad2/3 in the heart tissues and cultured cardiomyocytes and cardiac fibroblasts. In vitro experiments demonstrated that Ang II-induced cardiomyocyte hypertrophy and proliferation and activation of cardiac fibroblasts were markedly inhibited by klotho, LY364947 or the miR-132 inhibitor anti-miR-132. Both klotho and the TGF-β1 inhibitor LY364947 downregulated the miR-132 expression. Additionally, klotho decreased Ang II-induced protein expressions of cardiac fibroblast growth factor (FGF)23 in vivo and in vitro. The decreased protein levels of klotho in serum and renal tissues of Ang II-infused mice were elevated by klotho. Klotho downregulated the protein levels of TGF-β1 in renal tissues of Ang II-infused mice. In conclusion, our results suggest that klotho prevents Ang II-induced cardiac remodeling and dysfunction through modifying the TGF-β1-miR-132 axis, providing an experimental basis for clinical treatment on cardiac remodeling.