Merck
  • Slc7a5 regulates Kv1.2 channels and modifies functional outcomes of epilepsy-linked channel mutations.
Slc7a5 regulates Kv1.2 channels and modifies functional outcomes of epilepsy-linked channel mutations.
Nature communications (2018-10-26)
Victoria A Baronas, Runying Y Yang, Luis Carlos Morales, Simonetta Sipione, Harley T Kurata
ABSTRACT
Kv1.2 is a prominent voltage-gated potassium channel that influences action potential generation and propagation in the central nervous system. We explored multi-protein complexes containing Kv1.2 using mass spectrometry followed by screening for effects on Kv1.2. We report that Slc7a5, a neutral amino acid transporter, has a profound impact on Kv1.2. Co-expression with Slc7a5 reduces total Kv1.2 protein, and dramatically hyperpolarizes the voltage-dependence of activation by -47 mV. These effects are attenuated by expression of Slc3a2, a known binding partner of Slc7a5. The profound Slc7a5-mediated current suppression is partly explained by a combination of gating effects including accelerated inactivation and a hyperpolarizing shift of channel activation, causing channels to accumulate in a non-conducting state. Two recently reported Slc7a5 mutations linked to neurodevelopmental delay exhibit a localization defect and have attenuated effects on Kv1.2. In addition, epilepsy-linked gain-of-function Kv1.2 mutants exhibit enhanced sensitivity to Slc7a5.
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 antibody produced in mouse, clone NE14, ascites fluid
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution