Merck
  • Tumor-derived exosomes confer antigen-specific immunosuppression in a murine delayed-type hypersensitivity model.
Tumor-derived exosomes confer antigen-specific immunosuppression in a murine delayed-type hypersensitivity model.
PloS one (2011-08-11)
Chenjie Yang, Seon-Hee Kim, Nicole R Bianco, Paul D Robbins
ABSTRACT
Exosomes are endosome-derived small membrane vesicles that are secreted by most cell types including tumor cells. Tumor-derived exosomes usually contain tumor antigens and have been used as a source of tumor antigens to stimulate anti-tumor immune responses. However, many reports also suggest that tumor-derived exosomes can facilitate tumor immune evasion through different mechanisms, most of which are antigen-independent. In the present study we used a mouse model of delayed-type hypersensitivity (DTH) and demonstrated that local administration of tumor-derived exosomes carrying the model antigen chicken ovalbumin (OVA) resulted in the suppression of DTH response in an antigen-specific manner. Analysis of exosome trafficking demonstrated that following local injection, tumor-derived exosomes were internalized by CD11c+ cells and transported to the draining LN. Exosome-mediated DTH suppression is associated with increased mRNA levels of TGF-β1 and IL-4 in the draining LN. The tumor-derived exosomes examined were also found to inhibit DC maturation. Taken together, our results suggest a role for tumor-derived exosomes in inducing tumor antigen-specific immunosuppression, possibly by modulating the function of APCs.
Sigma-Aldrich
Monoclonal Anti-Chicken Egg Albumin (Ovalbumin) antibody produced in mouse, clone OVA-14, ascites fluid