Merck
Structural organization of lipid-functionalized-Au nanoparticles.
Colloids and surfaces. B, Biointerfaces (2018-05-08)
Alessandra Luchini, Gerardino D'Errico, Serena Leone, Zahra Vaezi, Annalisa Bortolotti, Lorenzo Stella, Giuseppe Vitiello, Luigi Paduano
ABSTRACT
Gold nanoparticles (AuNPs) are considered suitable systems for drug delivery and diagnostics with several applications in biomedicine. Size, shape and surface functionalization of these nanoparticles are important parameters influencing their behavior in a biological environment. This study describes the preparation and the characterization of lysophosphocholine coated AuNPs by means of Small Angle Neutron Scattering (SANS), Electron Paramagnetic Resonance (EPR) and Fluorescence Spectroscopy. In particular the structure of the functionalized AuNP suspension, as well as the physical properties, of the nanoparticle organic coating are discussed. The experimental results indicated that functionalized lysophosphocholine-AuNPs form aggregates, which are composed by nanoparticles with core-shell structure. Nevertheless, the nanoparticle suspension resulted to be stable, without significant structural rearrangements even when the temperature was increased to 50 °C. At the same time, experimental evidences also suggested that the 18LPC layer around AuNPs presented a reduced chain packing compared to pure 18LPC aggregates.
Sigma-Aldrich
6-Dodecanoyl-N,N-dimethyl-2-naphthylamine, suitable for fluorescence, ≥97.0% (HPLC)