Merck
  • Tandem Histone-Binding Domains Enhance the Activity of a Synthetic Chromatin Effector.
Tandem Histone-Binding Domains Enhance the Activity of a Synthetic Chromatin Effector.
ACS synthetic biology (2018-02-13)
Stefan J Tekel, Daniel A Vargas, Lusheng Song, Joshua LaBaer, Michael R Caplan, Karmella A Haynes
ABSTRACT
Fusion proteins that specifically interact with biochemical marks on chromosomes represent a new class of synthetic transcriptional regulators that decode cell state information rather than DNA sequences. In multicellular organisms, information relevant to cell state, tissue identity, and oncogenesis is often encoded as biochemical modifications of histones, which are bound to DNA in eukaryotic nuclei and regulate gene expression states. We have previously reported the development and validation of the "polycomb-based transcription factor" (PcTF), a fusion protein that recognizes histone modifications through a protein-protein interaction between its polycomb chromodomain (PCD) motif and trimethylated lysine 27 of histone H3 (H3K27me3) at genomic sites. We demonstrated that PcTF activates genes at methyl-histone-enriched loci in cancer-derived cell lines. However, PcTF induces modest activation of a methyl-histone associated reporter compared to a DNA-binding activator. Therefore, we modified PcTF to enhance its binding avidity. Here, we demonstrate the activity of a modified regulator called Pc2TF, which has two tandem copies of the H3K27me3-binding PCD at the N-terminus. Pc2TF has a smaller apparent dissociation constant value in vitro and shows enhanced gene activation in HEK293 cells compared to PcTF. These results provide compelling evidence that the intrinsic histone-binding activity of the PCD motif can be used to tune the activity of synthetic histone-binding transcriptional regulators.
Sigma-Aldrich
Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt, ≥95% (H-NMR), powder
Roche
Rapid DNA Ligation Kit
Sigma-Aldrich
Rabbit Anti-Chicken IgG Antibody, HRP conjugate, 0.8 mg/mL, Chemicon®