
Umckalin

Synonym(s):
5-Methoxyscopoletin, 7-Hydroxy-5, 6-dimethoxycoumarin, 7-Hydroxy-5,6-dimethoxy-2H-1-benzopyran-2-one
Empirical Formula (Hill Notation):
C11H10O5
CAS Number:
Molecular Weight:
222.19
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing