(S)-(−)-N-(Trifluoroacetyl)pyrrolidine-2-carbonyl chloride solution
Synonym(s):
(S)-(−)-N-(Trifluoroacetyl)prolyl chloride solution, TPC
Empirical Formula (Hill Notation):
C7H7ClF3NO2
CAS Number:
Molecular Weight:
229.58
Products
Product Category
Brand
Formula Weight
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing