Skip to Content
Merck
Silicon tetrachloride for synthesis

Silicon tetrachloride

Silicon tetrachloride for synthesis
Synonym(s):
STC, Tetrachlorosilane, Silicon tetrachloride
Linear Formula:
SiCl4
CAS Number:
Molecular Weight:
169.90

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Core