
Propoxycarbazone sodium salt

Synonym(s):
2-[(4-Methyl-5-oxo-3-propoxy-1,2,4-triazolin-1-yl)carbamidosulfonyl]benzoic acid methyl ester sodium salt
Empirical Formula (Hill Notation):
C15H17N4NaO7S
CAS Number:
Molecular Weight:
420.37
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing