Skip to Content
LB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Olmesartan medoxomil
Synonym(s):
Olmesartan medoxomil
Empirical Formula (Hill Notation):
C
29
H
30
N
6
O
6
CAS Number:
144689-63-4
Molecular Weight:
558.59
Products
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
SML1391
Olmesartan medoxomil,
≥98% (HPLC)
Expand
View Pricing
1478367
Olmesartan medoxomil,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
PHR1851
Olmesartan medoxomil,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
Y0001405
Olmesartan medoxomil,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
BP1173
Olmesartan medoxomil,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing