Skip to Content
Merck
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse clone 29.1, ascites fluid

Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse

Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse clone 29.1, ascites fluid
Synonym(s):
Anti-EGFR

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique