
Fmoc-D-Val-OH

Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-D-valine, Fmoc-D-valine, Fmoc-D-Val-OH
Empirical Formula (Hill Notation):
C20H21NO4
CAS Number:
Molecular Weight:
339.39
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing