Skip to Content
Merck
Ethyl levulinate ≥98%, FCC, FG

Ethyl levulinate

Ethyl levulinate ≥98%, FCC, FG
Synonym(s):
Ethyl 4-oxopentanoate
Linear Formula:
CH3COCH2CH2COOC2H5
CAS Number:
Molecular Weight:
144.17
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing