Skip to Content
Merck
Dinotefuran PESTANAL®, analytical standard

Dinotefuran

Dinotefuran PESTANAL®, analytical standard
Synonym(s):
(RS)-N-Methyl-N′-nitro-N″-[(tetrahydro-3-furanyl)methyl]guanidine
Empirical Formula (Hill Notation):
C7H14N4O3
CAS Number:
Molecular Weight:
202.21

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique