
Chlortoluron

Synonym(s):
3-(3-Chloro-4-methyl)-1,1-dimethylurea
Empirical Formula (Hill Notation):
C10H13ClN2O
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing