cataCXium A Pd G4 ChemBeads
Synonym(s):
cataCXium A Pd G4
Empirical Formula (Hill Notation):
C38H54NO3PPdS
CAS Number:
Molecular Weight:
742.30
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing