Skip to Content
Merck
Buprenorphine Related Compound A certified reference material, pharmaceutical secondary standard

Buprenorphine Related Compound A

Buprenorphine Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
N-(3-Butenyl)norbuprenorphine, (αS,5α,7α)-17-(3-Buten-1-yl)-α-(1,1-dimethylethyl)-4,5-epoxy-18,19-dihydro-3-hydroxy-6-methoxy-α-methyl-6,14-ethenomorphinan-7-methanol
Empirical Formula (Hill Notation):
C29H41NO4
CAS Number:
Molecular Weight:
467.64

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade