
Anti-SELP antibody produced in rabbit

Synonym(s):
Anti-CD62P antigen antibody produced in rabbit, Anti-GMP-140 antibody produced in rabbit, Anti-Granule membrane protein 140 antibody produced in rabbit, Anti-LECAM3 antibody produced in rabbit, Anti-Leukocyte-endothelial cell adhesion molecule 3 antibody produced in rabbit, Anti-P-selectin precursor antibody produced in rabbit, Anti-PADGEM antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing