Skip to Content
Merck
1-Z-Piperazine 98%

1-Z-Piperazine

1-Z-Piperazine 98%
Synonym(s):
1-(Benzyloxycarbonyl)piperazine, 1-Cbz-Piperazine, Benzyl piperazine-1-carboxylate
Empirical Formula (Hill Notation):
C12H16N2O2
CAS Number:
Molecular Weight:
220.27

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing