Merck
Effect of the concentration of the dopants (Er
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-06-02)
Ajay Kumar Choudhary, A Dwivedi, A Bahadur, S B Rai
ABSTRACT
Er
Sigma-Aldrich
Strontium oxide, 99.9% trace metals basis